missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FAM156A Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 16914-1-AP-150UL
Informations supplémentaires : Weight : 0.00010kg
Spécification
FAM156A | |
Polyclonal | |
Unconjugated | |
FAM156A | |
FAM156A, FAM156B, PRO0659, Protein FAM156A, TMEM29, TMEM29B, Transmembrane protein 29 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
29057 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q8NDB6 | |
FAM156A | |
FAM156A Fusion Protein Ag10001 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |