Learn More
FAM170A Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA558158
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: VSLSSYSSYK TCVSSLCVNK EERGMKIYYM QVQMNKGVAV SWETEETLES LEKQPRMEEV TLSEVVRVGT PPSDVSTRN Highest antigen sequence identity to the following orthologs: Mouse - 81%, Rat - 82%.
Acts as a nuclear transcription factor that positively regulates the expression of heat shock genes. Binds to heat shock promoter elements (HSE).Specifications
FAM170A | |
Polyclonal | |
Unconjugated | |
FAM170A | |
FAM170A; family with sequence similarity 170 member A; family with sequence similarity 170, member A; protein FAM170A; Zinc finger domain-containing protein; zinc finger protein ZNFD; Znf domain containing protein; ZNFD | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
340069 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry, Western Blot | |
0.4 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
A1A519 | |
FAM170A | |
Recombinant protein corresponding to Human FAM170A | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only