missing translation for 'onlineSavingsMsg'
Learn More

FAM185A Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 20911-1-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16840495

  • 4721.67 SEK / 150µL
Estimated Shipment: 06-12-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

FAM185A
Polyclonal
Unconjugated
FAM185A
FAM185A, Protein FAM185A
Rabbit
Antigen Affinity Chromatography
RUO
222234
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.25 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q8N0U4
FAM185A
FAM185A Fusion Protein Ag15041
150 μL
Primary
Human
Antibody
IgG
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers