missing translation for 'onlineSavingsMsg'
Learn More

FAM19A5 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 13948-1-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16882384

  • 3313.85 SEK / 150µL
Estimated Shipment: 08-12-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

FAM19A5
0.38 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q7Z5A7, Q91WE9
Chemokine like protein TAFA 5, FAM19A5, Protein FAM19A5, QLLK5208, TAFA 5, TAFA5, UNQ5208
Rabbit
IgG
150 μL
-20°C
Primary
106014, 25817, 500915
Immunohistochemistry (Paraffin)
Unconjugated
FAM19A5
Liquid
Fam19a5
FAM19A5 Fusion Protein Ag5058
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Videos
SDS
Documents
Certificates
Special Offers

Special Offers