missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FAM20C Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | FAM20C |
---|---|
Concentration | 0.36 mg/mL |
Applications | Immunohistochemistry (Paraffin), Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16823975
|
Proteintech
25395-1-AP-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 24-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16813975
|
Proteintech
25395-1-AP-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 24-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Calcium-binding protein which may play a role in dentin mineralization.Specifications
FAM20C | |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
Q5MJS3, Q8IXL6 | |
304334, 56975, 80752 | |
FAM20C Fusion Protein Ag22061 | |
Primary | |
-20°C | |
FAM20C |
0.36 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Dentin matrix protein 4, DMP 4, DMP4, FAM20C, Protein FAM20C, RNS | |
FAM20C | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |