Learn More
FAM210B Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA560066
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: RLDARLLRTA RGDCRGHQDP SQATGTTGSS VSCTEEKKQS KSQQLK Highest antigen sequence identity to the following orthologs: Mouse - 59%, Rat - 61%.
FAM210B Polyclonal antibody specifically detects FAM210B in Human samples. It is validated for Immunohistochemistry.Specifications
FAM210B | |
Polyclonal | |
Unconjugated | |
FAM210B | |
2010011I20Rik; 5A3; AI430961; AV108736; C20orf108; dJ1167H4.1; FAM210B; family with sequence similarity 210 member B; family with sequence similarity 210, member B; protein FAM210B; Protein FAM210B, mitochondrial; PSEC0265; RGD1311378; transmembrane protein C20orf108; transmembrane protein C20orf108 homolog | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
116151 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q96KR6 | |
FAM210B | |
Recombinant protein corresponding to Human FAM210B | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only