missing translation for 'onlineSavingsMsg'
Learn More

FAM214A Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Brand:  Thermo Scientific PA559092

Additional Details : Weight : 0.02000kg

Product Code. 15706961

  • 4575.00 SEK / 100µL
Estimated Shipment: 26-04-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Immunogen sequence: EVLRTTLKHS NVWRKHNFHS LDGTSTRAFH PQTGLPLLSS PVPQRKTQSG CFDLDSSLLH LKSFSSRSPR PCLNIEDDPD IHEKPFLSSS APPI Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 87%.

FAM214A Polyclonal antibody specifically detects FAM214A in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin).
Specifications

Specifications

FAM214A
Polyclonal
Unconjugated
FAM214A
FAM214A; family with sequence similarity 214 member A; family with sequence similarity 214, member A; KIAA1370; Protein FAM214A
Rabbit
Antigen affinity chromatography
RUO
56204
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunocytochemistry, Immunohistochemistry (Paraffin)
Lot-Specific
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q32MH5
FAM214A
Recombinant protein corresponding to Human FAM214A
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Research Use Only