Learn More
FAM214B Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA561822
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: PSQPPVRQGA LQGGLLMGYS PAGGATSPGV YQVSIFSPPA GTSEPHRALK RQAPSTEGPR ELKRGPGLGA REGLPPEEPS TV Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 93%.
Specifications
FAM214B | |
Polyclonal | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q7L5A3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
80256 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry | |
Unconjugated | |
FAM214B | |
FAM214B | |
Recombit protein corresponding to Human FAM214B | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only