Learn More
FAM217A Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA557277
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: SVDKQVGPYP GLPMPLGLCW PYADGDFFKN RNEIHVSSCS TIENNDGETL PAPNWNLKHG NSSVEENFTD ESDLSENEKT NDTLLSYFKK VDLNLKPETI K Highest antigen sequence identity to the following orthologs: Mouse - 73%, Rat - 70%.
FAM217A Polyclonal antibody specifically detects FAM217A in Human samples. It is validated for Immunohistochemistry.Specifications
FAM217A | |
Polyclonal | |
Unconjugated | |
FAM217A | |
1700026J04Rik; C35H6orf146; C6orf146; Fam217a; family with sequence similarity 217 member A; family with sequence similarity 217, member A; LOW QUALITY PROTEIN: protein FAM217A; protein FAM217A; uncharacterized protein C6orf146; uncharacterized protein C6orf146 homolog | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
222826 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8IXS0 | |
FAM217A | |
Recombinant protein corresponding to Human FAM217A | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only