missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FAM221B Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA555349
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: IIEEPHITMD AEKHPPSKDP SAEDLQENHI SESFLKPSTS ETPLEPHTSE SPLVPSPSQI PLEAHSPETH QEPSISETP Highest antigen sequence identity to the following orthologs: Mouse - 34%, Rat - 38%.
FAM221B Polyclonal antibody specifically detects FAM221B in Human samples. It is validated for Immunohistochemistry.Specifications
FAM221B | |
Polyclonal | |
Unconjugated | |
FAM221B | |
C9orf128; FAM221B; family with sequence similarity 221 member B; family with sequence similarity 221, member B; protein FAM221B; uncharacterized protein C9orf128 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
392307 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.4 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
A6H8Z2 | |
FAM221B | |
Recombinant protein corresponding to Human FAM221B | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only