Learn More
FAM43A Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA561718
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: SDMKAELSQL ISDLGELSFG NDVRTLQADL RVTRLLSGDS TGSESSIEGG GPDATSATAG DSSRQADGAS ADEP Highest antigen sequence identity to the following orthologs: Mouse - 76%, Rat - 78%.
FAM43A Polyclonal antibody specifically detects FAM43A in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin).Specifications
FAM43A | |
Polyclonal | |
Unconjugated | |
FAM43A | |
Fam43a; family with sequence similarity 43 member A; family with sequence similarity 43, member A; PP7298; Protein FAM43A; somitovasculin; Tuf1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
131583 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin) | |
0.5 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8N2R8 | |
FAM43A | |
Recombinant protein corresponding to Human FAM43A | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only