missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Invitrogen™ FAM48A Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA558586
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
Immunogen sequence: DSETIRLPYE EGELLEYLDA EELPPILVDL LEKSQVNIFH CGCVIAEIRD YRQSSNMKSP GYQSRHILLR PTMQTLICDV HSITSDNHKW TQED Highest antigen sequence identity to the following orthologs: Mouse - 99%, Rat - 99%.
Required for MAP kinase p38 (MAPK11, MAPK12, MAPK13 and/or MAPK14) activation during gastrulation. Required for down-regulation of E-cadherin during gastrulation by regulating E-cadherin protein level downstream from NCK-interacting kinase (NIK) and independently of the regulation of transcription by FGF signaling and Snail. Required for starvation-induced ATG9A trafficking during autophagy. [UniProt].
Specifikationer
| FAM48A | |
| Polyclonal | |
| Unconjugated | |
| SUPT20H | |
| bA421P11.4; C13; C13orf19; Fam48a; family with sequence similarity 48, member A; FP757; p38-interacting protein; P38IP; protein FAM48A; RGD1307812; SPT20; SPT20 homolog, SAGA complex component; suppressor of Ty 20; suppressor of Ty 20 homolog; Supt20; SUPT20H; transcription factor (p38 interacting protein); transcription factor SPT20 homolog; tumor rejection antigen | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 55578 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8NEM7 | |
| SUPT20H | |
| Recombinant protein corresponding to Human FAM48A. Recombinant protein control fragment (Product #RP-97915). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering