missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FAM58A Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA567796
Additional Details : Weight : 0.01000kg
Description
Predicted to react with Mouse and Rat samples.
FAM58A Polyclonal antibody specifically detects FAM58A in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
FAM58A | |
Polyclonal | |
Unconjugated | |
CCNQ | |
Ccnq; CDK10-activating cyclin; cyclin M; cyclin Q; cyclin-M; Cyclin-Q; Cyclin-related protein FAM58A; Fam58a; Fam58b; family with sequence similarity 58 member A; family with sequence similarity 58, member A; family with sequence similarity 58, member B; RGD1305651; STAR | |
Rabbit | |
affinity chromatography | |
RUO | |
303321, 92002 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q4QQW5, Q8N1B3 | |
CCNQ | |
Synthesized peptide derived from human FAM58A. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |