Learn More
FAM69B Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA561338
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: FVEELFHGSY GTFYMCETTL ANVGYTATYD FKMADLQQVA PEATVRRFLQ GRRCEHSTDC TYGRDCRAPC DRLMRQCKGD LIQPNLAKVC ALLRGYL Highest antigen sequence identity to the following orthologs: Mouse - 94%, Rat - 93%.
FAM69B Polyclonal antibody specifically detects FAM69B in Human samples. It is validated for Immunohistochemistry.Specifications
FAM69B | |
Polyclonal | |
Unconjugated | |
FAM69B | |
AB030186; AW209000; B230317C12Rik; C9orf136; DIPK1B; Divergent protein kinase domain 1B; Fam69b; family with sequence similarity 69 member B; family with sequence similarity 69, member B; pancreatitis-induced protein 49; PIP49; pp6977; Protein FAM69B | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
138311 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q5VUD6 | |
FAM69B | |
Recombinant protein corresponding to Human FAM69B | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only