Learn More
FAM71E2 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 20904-1-AP-150UL
Additional Details : Weight : 0.00010kg
Specifications
FAM71E2 | |
Polyclonal | |
Unconjugated | |
FAM71E2 | |
C19orf16, FAM71E2, Protein FAM71E2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
284418 | |
-20°C | |
Liquid |
Western Blot, Immunohistochemistry (Paraffin) | |
0.54 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q8N5Q1 | |
FAM71E2 | |
FAM71E2 Fusion Protein Ag15027 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |