missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FAM89B Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA550410
Additional Details : Weight : 0.01000kg
Description
The antibody detects endogenous levels of total FAM89B protein.
FAM89B Polyclonal antibody specifically detects FAM89B in Human samples. It is validated for Immunohistochemistry (Paraffin).Specifications
FAM89B | |
Polyclonal | |
Unconjugated | |
Fam89b | |
1110021A21Rik; C184L ORF2 protein; C184m; C184M protein; FAM89B; family with sequence similarity 89 member B; family with sequence similarity 89, member B; Leucine repeat adapter protein 25; leucine repeat adaptor protein 25; Lrap25; Mammary tumor virus receptor 2; mammary tumor virus receptor homolog 1; MMTV; MMTV receptor; MTVR; MTVR1; Mtvr2; protein FAM89B | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
23625 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
Q8N5H3 | |
Fam89b | |
Full length fusion protein of Human FAM89B | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |