Learn More
FAM92A1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA557048
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: KTIFSEFITI EMLFHGKALE VYTAAYQNIQ NIDEDEDLEV FRNSLYAPDY SSRLDIVRAN SKSPLQRSLS AKCVSGTGQE AGWAATCQTL I Highest antigen sequence identity to the following orthologs: Mouse - 81%, Rat - 80%.
Acts as a positive regulator of ciliary hedgehog signaling. Probable regulator of ciliogenesis involved in limb morphogenesis. In cooperation with CBY1 it is involved in the recruitment and fusion of endosomal vesicles at distal appendages during early stages of ciliogenesis. [UniProt]Specifications
FAM92A1 | |
Polyclonal | |
Unconjugated | |
FAM92A | |
6720467C03Rik; Fam92a; FAM92A1; family with sequence similarity 92 member A1; family with sequence similarity 92, member A; family with sequence similarity 92, member A1; Protein FAM92A; protein FAM92A1; RGD1310681 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
137392 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Western Blot | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
A1XBS5 | |
FAM92A | |
Recombinant protein corresponding to Human FAM92A1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only