Learn More
Famciclovir-d4, TRC
Product Code. 30394705
Change view
Click to view available options
Kvantitet:
2.5 mg
25 mg
Unit Size:
2.5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet unitSize
30394705 25 mg -
30396791 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30394705 Supplier Toronto Research Chemicals Supplier No. TRCF10112725MG
 Begränsad artikel
Beräknad leverans: 14-08-2026
Add to basket
Add to basket
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Högrena organiska molekyler och analytiska standarder, strategiskt levererade världen över för att möjliggöra innovation och kommersiell framgång.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1020719-42-9
Molekylformel C14 2H4 H15 N5 O4
Molekylvikt (g/mol) 325.36
IUPAC-namn [2-[acetyloxi(dideuterio)metyl]-4-(2-aminopurin-9-yl)-1,1-dideuteriobutyl] acetat
LEDER [2H]C([2H])(OC(=O)C)C(CCn1cnc2cnc(N)nc12)C([2H])([2H])OC(=O)C
Specifications
Kemiskt namn eller material Famciclovir-d4
Kvantitet 25 mg
InChI formel InChI=1S/C14H19N5O4/c1-9(20)22-6-11(7-23-10(2)21)3-4-19-8-17-12-5-16-14(15)18-13(12)19/h5,8,11H,3-4,6-7H2,1-2H3,(H2,15,16,18)/i6D2,7D2
Formel vikt 325.1688