Läs mer
Famotidine-13C3, TRC
Produktkod. 30133881
Change view
Kvantitet:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30133881 10 mg
17983595 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30133881 Leverantör Toronto Research Chemicals Leverantörsnummer TRCF10225210MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1185241-48-8
Molekylformel C513C3H15N7O2S3
Molekylvikt (g/mol) 340.42
Synonym 3-[[[2-[(Aminoiminomethyl)amino]-4-thiazolyl]methyl]thio]-N-(aminosulfonyl)propanimidamide-13C3, N-(Aminosulfonyl)-3-[[[2-[(diaminomethylene)amino]-4-thiazolyl]methyl] thio]propanimidamide-13C3, Amfamox-13C3, Antodine-13C3, Famodin-13C3, Famosan-13C3, Ganor-13C3, Lecedil-13C3, Mosul-13C3, Motiax-13C3, Ulfamid-13C3;
LEDER NS(/N=C(N)/CCSC[13C]1=CS[13C](/N=[13C](N)/N)=N1)(=O)=O
Specifikationer
Kemiskt namn eller material Famotidine-13C3
Kvantitet 10 mg
InChI formel InChI=1S/C8H15N7O2S3/c9-6(15-20(12,16)17)1-2-18-3-5-4-19-8(13-5)14-7(10)11/h4H,1-3H2,(H2,9,15)(H2,12,16,17)(H4,10,11,13,14)