Läs mer
Famotidine Acid Methyl Ester Hydrochloride (Famotidine Impurity), TRC
Produktkod. 30138411
Change view
Kvantitet:
100 mg
250 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30138411 250 mg
30136291 1 g
30009510 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30138411 Leverantör Toronto Research Chemicals Leverantörsnummer TRCF102295250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1798006-30-0
Molekylformel C9 H14 N4 O2 S2 . Cl H
Molekylvikt (g/mol) 310.82
Synonym Propanoic acid, 3-[[[2-[(aminoiminomethyl)amino]-4-thiazolyl]methyl]thio]-, methyl ester, hydrochloride (1:1), Methyl 3-[[[2-[(aminoiminomethyl)amino]-4-thiazolyl]methyl]thio]propanoate hydrochloride, Methyl 3-[[[2-[(Diaminomethylidene)amino]thiazol-4-yl]methyl]sulfanyl]propanoate hydrochloride
IUPAC-namn methyl 3-[[2-(diaminomethylideneamino)-1,3-thiazol-4-yl]methylsulfanyl]propanoate;hydrochloride
LEDER Cl.COC(=O)CCSCc1csc(N=C(N)N)n1
Specifikationer
Kemiskt namn eller material Famotidine acid methyl ester hydrochloride
Anteckningar om renhetsgrad HPLC
Kvantitet 250 mg
InChI formel InChI=1S/C9H14N4O2S2.ClH/c1-15-7(14)2-3-16-4-6-5-17-9(12-6)13-8(10)11;/h5H,2-4H2,1H3,(H4,10,11,12,13);1H
Formel vikt 310.0325
Procent renhet >95%