Läs mer
Fanapanel hydrate, MedChemExpress
Produktkod. 16436308
Encompass_Preferred
Change view
Kvantitet:
10 mg
50 mg
Förpackningsstorlek:
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
16436308 10 mg 10mg
16432427 50 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 16436308 Leverantör Medchem Express Leverantörsnummer HY15069A
MedChemExpress Fanapanel hydrate (ZK200775 hydrate) is a highly selective AMPA/kainate antagonist with little activity against NMDA; have Ki values of 3.2 nM, 100 nM, and 8.5 μM against quisqualate, kainate, and NMDA, respectively.
Fanapanel hydrate (ZK200775 hydrate) is a highly selective AMPA/kainate antagonist with little activity against NMDA
Specifikationer
Kemiskt namn eller material Fanapanel hydrate
Fysisk form Solid
CAS 1255517-78-2
Färg Light Yellow
Kvantitet 10 mg
Anteckningar om renhetsgrad Research
Molekylformel C14H17F3N3O7P
För användning med (applikation) Neuroscience-Neurodegeneration
Synonym ZK200775 hydrate MPQX hydrate
Löslighetsinformation DMSO : 5 mg/mL (11.70 mM; Need ultrasonic)
LEDER O=C(N1CP(O)(O)=O)C(NC(C1=C2)=CC(C(F)(F)F)=C2N3CCOCC3)=O.[H]O[H]
Molekylvikt (g/mol) 427.27
Formel vikt 427.27
Procent renhet 99.66%