missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ FANCA (Human) Recombinant Protein
Human FANCA full-length ORF ( AAH08979, 1 a.a. - 297 a.a.) recombinant protein with GST-tag at N-terminal.
Brand: Abnova™ H00002175-P01.25ug
Additional Details : Weight : 0.00010kg
Description
- Sequence: MSDSWVPRSASGQDPGGRRRAWAELLAGRVKREKYNPERAQKLKESAVRLLRSHQDLNALLLEVEGPLCKKLSLSKVIDCDSSEAYANHSSSFIGSALQDQASRLGVPVGILSAGMVASSVGQICTAPAETSHPVLLTVEQRKKLSSLLEFARYLLAHSMFSRLSFCQELWKIQSSLLLEAVWHLHVQGIVSLQELLESHPDMHAVGSWLFRNLCCLCEQMEASCQHADVARAMLSDFVQMFVLRGFQKNSDLRRTVEPEKMPQVAVDVLQRMLIFALDALAAGVQEESSTHKIVRC
Specifications
AAH08979 | |
Fanconi anemia, complementation group A | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FANCA | |
GST |
2175 | |
Wheat germ expression system | |
25 μg | |
FA, FA-H, FA1, FAA, FACA, FAH, FANCH, MGC75158 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |