missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ FASN (Human) Recombinant Protein

Human FASN full-length ORF ( AAH07909, 1 a.a. - 439 a.a.) recombinant protein with GST-tag at N-terminal.

Brand:  Abnova™ H00002194-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16134861

  • 6150.00 SEK / 25µg
Estimated Shipment: 06-10-2022
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

  • Sequence: MSTNDTIVSGTLPQRMASCLEVLDLFLNQPHMVLSSFVLAEKAAAYRDRDSQRDLVEAVAHILGIRDLAAVNLDSSLADLGLDSLMSVEVRQTLERELNLVLSVREVRQLTLRKLQELSSKADEASELACPTPKEDGLAQQQTQLNLRSLLVNPEGPTLMRLNSVQSSERPLFLVHPIEGSTTVFHSLASGLSIPTYGLQCTRAAPLDSIHSLAAYYIDCIRQVQPEGPYRVAGYSYGACVAFEMCSQLQAQQSPAPTHNSLFLFDGSPTYVLAYTQSYRAKLTPGCEAEAETEAICFFVQQFTDMEHNRVLEALLPLKGLEERVAAAVDLIIKSHQGLDRQELSFAARSFYYKLRAAEQYTPKAKYHGNVMLLRAKTGGAYGEDLGADYNLSQVCDGKVSVHVIEGDHRTLLEGSGLESIISIIHSSLAEPRVSVREG
Specifications

Specifications

AAH07909
fatty acid synthase
125% SDS-PAGE Stained with Coomassie Blue
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FASN
GST
2194
Wheat germ expression system
25 μg
FAS, MGC14367, MGC15706, OA-519, SDR27X1
Wheat Germ (in vitro)
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer
Videos
SDS
Documents
Certificates
Special Offers

Special Offers