Läs mer
FASN-IN-1, MedChemExpress
Produktkod. 16477237
Encompass_Preferred
Change view
Kvantitet:
5 mg
10 mg
50 mg
100 mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
16477237 5 mg
16444528 10 mg
16435386 100 mg
16410837 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Produktkod. 16477237 Leverantör Medchem Express Leverantörsnummer HY111777
MedChemExpress FASN-IN-1 is a fatty acid synthase (FASN) inhibitor extracted from patent WO2015134790A1, compound 56.
FASN-IN-1 is a fatty acid synthase (FASN) inhibitor extracted from patent WO2015134790A1, compound 56.
Specifikationer
Kemiskt namn eller material FASN-IN-1
CAS 1808260-84-5
Färg Light Yellow
Anteckningar om renhetsgrad Research
Molekylformel C18H25N3O3S2
Kvantitet 5 mg
Löslighetsinformation DMSO : 100 mg/mL (252.82 mM; Need ultrasonic)
LEDER CC[C@H](C)C(NC1=NC(C2=CC=C(S(=O)(N(CC)CC)=O)C=C2)=CS1)=O
Molekylvikt (g/mol) 395.54
Formel vikt 395.54
Procent renhet 99.62%
Kvalitet Research
Fysisk form Solid
För användning med (applikation) Neuroscience-Neuromodulation