missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FASTKD1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA549834
Additional Details : Weight : 0.01000kg
Description
The antibody detects endogenous levels of total FASTKD1 protein.
FASTKD1 Polyclonal antibody specifically detects FASTKD1 in Human, Mouse samples. It is validated for Western Blot.Specifications
FASTKD1 | |
Polyclonal | |
Unconjugated | |
FASTKD1 | |
5330408N05Rik; FAST kinase domain-containing protein 1; FAST kinase domain-containing protein 1, mitochondrial; FAST kinase domains 1; Fastkd1; KIAA1800; mKIAA1800; RGD1563531 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
320720, 79675 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q53R41, Q6DI86 | |
FASTKD1 | |
Synthesized peptide derived from internal of human FAKD1. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |