Läs mer
Fasudil monohydrochloride, 99+%
Produktkod. 15453469
Change view
Kvantitet:
250 mg
500 mg
1 g
Förpackningsstorlek:
1g
250mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet Förpackningsstorlek
15453469 500 mg 500mg
15443469 250 mg 250mg
15433469 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 15453469 Leverantör Thermo Scientific Alfa Aesar Leverantörsnummer J60751.ME
 Begränsad artikel
Beräknad leverans: 14-10-2026
CAS: 105628-07-7 | C14H18ClN3O2S | 327.827 g/mol
Fasudil monohydrochloride is used in the prevention and treating cardiovascular and cerebrovascular diseases. It is also used in the inhibition of proliferation of vascular smooth muscle cells. It serves as a selective Rho-associated protein kinase (ROCK) inhibitor. Further, it protects against methyl mercury-induced axonal degeneration.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Solubility
Soluble in water and dimethyl sulfoxide. Insoluble in ethanol.
Notes
Incompatible with acids, bases and oxidizing agents.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 105628-07-7
Molekylformel C14H18ClN3O2S
Molekylvikt (g/mol) 327.827
MDL-nummer MFCD00943198
InChI-nyckel LFVPBERIVUNMGV-UHFFFAOYSA-N
Synonym fasudil hydrochloride, fasudil hcl, eril, at 877 hydrochloride, fasudil monohydrochloride, ha 1077 hydrochloride, unii-sq04n8s7br, 5-1,4-diazepan-1-yl sulfonyl isoquinoline hydrochloride, sq04n8s7br
PubChem CID 163751
IUPAC-namn 5-(1,4-diazepan-1-ylsulfonyl)isoquinoline;hydrochloride
LEDER C1CNCCN(C1)S(=O)(=O)C2=CC=CC3=C2C=CN=C3.Cl
Specifikationer
Kemiskt namn eller material Fasudil monohydrochloride
Färg White
Kvantitet 500 mg
Merck Index 14,3942
Löslighetsinformation Soluble in water and dimethyl sulfoxide. Insoluble in ethanol.
Formel vikt 327.83
Procent renhet ≥99%
Fysisk form Powder