missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ FATE1 (Human) Recombinant Protein
Human FATE1 full-length ORF (BAB71454.1, 1 a.a. - 183 a.a.) recombinant protein with GST tag at N-terminal.
Brand: Abnova™ H00089885-P01.25ug
Additional Details : Weight : 0.00010kg
Description
- Sequence: MAGGPPNTKAEMEMSLAEELNHGRQGENQEHLVIAEMMELGSRSRGASQKKQKLEQKAAGSASAKRVWNMTATRPKKMGSQLPKPRMLRESGHGDAHLQEYAGNFQGIRFHYDRNPGTDAVAQTSLEEFNVLEMEVMRRQLYAVNRRLRALEEQGATWRHRETLIIAVLVSASIANLWLWMNQ
Specifications
BAB71454.1 | |
fetal and adult testis expressed 1 | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FATE1 | |
GST |
89885 | |
Wheat germ expression system | |
25 μg | |
FATE | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |