Läs mer
Tocris Bioscience™ Fatostatin A
Produktkod. 18015332
Change view
Kvantitet:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
18015332 10 mg
18054542 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 18015332 Leverantör Tocris Bioscience™ Leverantörsnummer 4444/10
Cell permeable inhibitor of SREBP activation
Inhibitor of sterol regulatory element binding protein (SREBP); impairs the activation of SREBP-1 and SREBP-2. Exhibits antiproliferative effects in DU 145 cells independently of IGF-1 signaling (IC50 = 0.1 μM); reverses hyperglycemia in diabetic (ob/ob) mice. Cell permeable.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 298197-04-3
Molekylformel C18H19BrN2S
Specifikationer
Kemiskt namn eller material Fatostatin A
Kvantitet 10 mg
Formel vikt 375.33
Renhet >97%
Molekylvikt (g/mol) 375.328