missing translation for 'onlineSavingsMsg'
Learn More

FATP4 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 11013-1-AP-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16827793

  • 1236.20 SEK / 20µL
Estimated Shipment: 03-12-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This gene encodes a member of a family of fatty acid transport proteins, which are involved in translocation of long-chain fatty acids cross the plasma membrane. This protein is expressed at high levels on the apical side of mature enterocytes in the small intestine, and appears to be the principal fatty acid transporter in enterocytes. Clinical studies suggest this gene as a candidate gene for the insulin resistance syndrome. Mutations in this gene have been associated with ichthyosis prematurity syndrome.
Specifications

Specifications

FATP4
0.13 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q6P1M0
ACSVL4, FATP 4, FATP4, Fatty acid transport protein 4, SLC27A4
Rabbit
IgG
20 μL
-20°C
Primary
10999
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Unconjugated
SLC27A4
Liquid
SLC27A4
FATP4 Fusion Protein Ag1490
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human
Videos
SDS
Documents
Certificates
Special Offers

Special Offers