Learn More
Abcam Fatty Acid Amide Hydrolase Activity Assay Kit (Fluorometric)
Product Code. 32117778 Shop All Abcam Products
Change view
Click to view available options
100 Tests
Product Code.
32117778 100 Tests
1 options
Product Code. 32117778 Supplier Abcam Supplier No. ab252895100Test
 Restricted Item
Beräknad leverans: 25-11-2026
Measure FAAH1 activity in Cell Samples, Other samples, Cell/Tissue Extracts with Fatty Acid Amide Hydrolase Activity Assay Kit AB252895. Sensitivity = 0.1 μU. Cited in 3 publications. Use with Microplate.
Measure FAAH1 activity in Cell Samples, Other samples, Cell/Tissue Extracts with Fatty Acid Amide Hydrolase Activity Assay Kit AB252895. Sensitivity = 0.1 μU. Cited in 3 publications. Use with Microplate.
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Quantity 100 Tests
Regulatory Status RUO