missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ FBLN1 (Human) Recombinant Protein
Human FBLN1 full-length ORF ( AAH22497.1, 1 a.a. - 683 a.a.) recombinant protein with GST-tag at N-terminal.
Brand: Abnova™ H00002192-P01.25ug
Additional Details : Weight : 0.00010kg
Description
- Sequence: MERAAPSRRVPLQLLLLGGLALLAAGVDADVLLEACCADGHRMATHQKDCSLPYATESKECRMVQEQCCHSQLEELHCATGISLANEQDRCATPHGDNASLEATFVKRCCHCCLLGRAAQAQGQSCEYSLMVGYQCGQVFRACCVKSQETGDLDVGGLQETDKIIEVEEEQEDPYLNDRCRGGGPCKQQCRDTGDEVVCSCFVGYQLLSDGVSCEDVNECITGSHSCRLGESCINTVGSFRCQRDSSCGTGYELTEDNSCKDIDECESGIHNCLPDFICQNTLGSFRCRPKLQCKSGFIQDALGNCIDINECLSISAPCPIGHTCINTEGSYTCQKNVPNCGRGYHLNEEGTRCVDVDECAPPAEPCGKGHRCVNSPGSFRCECKTGYYFDGISRMCVDVNECQRYPGRLCGHKCENTLGSYLCSCSVGFRLSVDGRSCEDINECSSSPCSQECANVYGSYQCYCRRGYQLSDVDGVTCEDIDECALPTGGHICSYRCINIPGSFQCSCPSSGYRLAPNGSNCQDIDECVTGIHNCSINETCFNIQGGFRCLAFECPENYRRSAATRCERLPCHENRECSKLPLRITYYHLSFPTNIQAPAVVFRMGPSSAVPGDSMQLAITGGNEEGFFTTRKVSPHSGVVALTKPVPEPRDLLLTVKMDLSRHGTVSSFVAKLFIFVSAEL
Specifications
AAH22497.1 | |
fibulin 1 | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FBLN1 | |
GST |
2192 | |
Wheat germ expression system | |
25 μg | |
FBLN | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |