Learn More
FBXO4 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA564893
Additional Details : Weight : 0.01000kg
Description
Immunogen sequence: HEWQDEFSHIM AMTDPAFGSS GRPLLVLSCI SQGDVKRMPC FYLAHELHLN LLNHPWLVQD TEAETLTGFL NGIEWILEEV ESK Highest antigen sequence identity to the following orthologs - mouse 94%, rat 93%.
This gene encodes a member of the F-box protein family which is characterized by an approximately 40 amino acid motif, the F-box. The F-box proteins constitute one of the four subunits of the ubiquitin protein ligase complex called SCFs (SKP1-cullin-F-box), which function in phosphorylation-dependent ubiquitination. The F-box proteins are divided into 3 classes: Fbws containing WD-40 domains, Fbls containing leucine-rich repeats, and Fbxs containing either different protein-protein interaction modules or no recognizable motifs. The protein encoded by this gene belongs to the Fbxs class. Alternative splicing of this gene generates 2 transcript variants.Specifications
FBXO4 | |
Polyclonal | |
Unconjugated | |
FBXO4 | |
1700096C12Rik; AI851261; AW494535; F-box only protein 4; F-box protein 4; F-box protein Fbx4; FBX4; Fbxo4 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
26272 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9UKT5 | |
FBXO4 | |
Recombinant Human FBXO4 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
For Research Use Only