Learn More
Abnova™ FBXO45 (Human) Recombinant Protein
Human FBXO45 partial ORF ( XP_117294.3, 39 a.a. - 148 a.a.) recombinant protein with GST-tag at N-terminal.
4515.00 SEK - 6845.00 SEK
Specifications
Accession Number | XP_117294.3 |
---|---|
Gene ID (Entrez) | 200933 |
Name | F-box protein 45 |
Preparation Method | Wheat germ expression system |
Quality Control Testing | 125% SDS-PAGE Stained with Coomassie Blue |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16132397
|
Abnova™
H00200933-Q01.10UG |
10 μg |
4515.00 SEK
10µg |
Estimated Shipment: 28-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16142397
|
Abnova™
H00200933-Q01.25UG |
25 μg |
6845.00 SEK
25µg |
Estimated Shipment: 28-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Sequence: LPSRVLELVFSYLELSELRSCALVCKHWYRCLHGDENSEVWRSLCARSLAEEALRTDILCNLPSYKAKIRAFQHAFSTNDCSRNVYIKKNGFTLHRNPIAQSTDGARTKI
Specifications
XP_117294.3 | |
F-box protein 45 | |
125% SDS-PAGE Stained with Coomassie Blue | |
Fbx45 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |
200933 | |
Wheat germ expression system | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FBXO45 | |
GST |