Learn More
Abnova™ FBXO45 (Human) Recombinant Protein
Human FBXO45 partial ORF ( XP_117294.3, 39 a.a. - 148 a.a.) recombinant protein with GST-tag at N-terminal.
Brand: Abnova™ H00200933-Q01.10ug
Additional Details : Weight : 0.00010kg
Description
- Sequence: LPSRVLELVFSYLELSELRSCALVCKHWYRCLHGDENSEVWRSLCARSLAEEALRTDILCNLPSYKAKIRAFQHAFSTNDCSRNVYIKKNGFTLHRNPIAQSTDGARTKI
Specifications
XP_117294.3 | |
F-box protein 45 | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FBXO45 | |
GST |
200933 | |
Wheat germ expression system | |
10 μg | |
Fbx45 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |