Läs mer
Fc gamma RIIB/CD32b Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-14905
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
Fc gamma RIIB/CD32b Polyclonal specifically detects Fc gamma RIIB/CD32b in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence.
Specifikationer
| Fc gamma RIIB/CD32b | |
| Polyclonal | |
| Unconjugated | |
| PBS (pH 7.0), 1.0% BSA and 20% Glycerol with 0.01% Thimerosal | |
| CD32 antigen, CD32B, CD32Fc fragment of IgG, low affinity II, receptor for (CD32), CDw32, Fc fragment of IgG, low affinity IIb, receptor (CD32), Fc fragment of IgG, low affinity IIb, receptor for (CD32), Fc gamma RIIb, FCG2, Fc-gamma RII-b, fc-gamma-RIIb, FCGR2, fcRII-b, IGFR2, IgG Fc receptor II-b, low affinity immunoglobulin gamma Fc region receptor II-b | |
| Rabbit | |
| 34 kDa | |
| 0.1 mL | |
| Immunology | |
| 2213 | |
| Human | |
| IgG |
| Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
| 1mg/mL | |
| Western Blot 1:500 - 1:3000, Flow Cytometry Assay-dependent dilution., Immunocytochemistry/Immunofluorescence 1:100 - 1:1000 | |
| P31994 | |
| FCGR2B | |
| Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human CD32B. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human. Predicted cross-reactivity based on sequence identity: Rhesus Monkey (92%). | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Korrigering av produktinnehåll
Produkttitel
For Research Use Only
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering