Läs mer
Fe(III) meso-Tetra(4-carboxyphenyl)porphine Chloride, TRC
Produktkod. 17918304
Change view
Kvantitet:
25 mg
100 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17918304 25 mg
17968974 100 mg
17908974 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 17918304 Leverantör Toronto Research Chemicals Leverantörsnummer TRCF32252825MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 55266-17-6
Molekylformel C48H28ClFeN4O8
Molekylvikt (g/mol) 880.06
Synonym Hydrogen (SP-5-12)-Chloro[[4,4',4' ',4' ' '-(21H,23H-porphine-5,10,15,20-tetrayl-κN21,κN22,κN23,κN24)tetrakis[benzoato]](6-)]ferrate(4-), 4,4',4' ',4' ' '-(21H,23H-Porphine-5,10,15,20-tetrayl)tetrakisbenzoic Acid Iron Complex, (SP-5-12)-Chloro[[4,4',4' ',4' ' '-(21H,23H-porphine-5,10,15,20-tetrayl-κN21,κN22,κN23,κN24)tetrakis[benzoato]](6-)]Ferrate(4-) Tetrahydrogen;
LEDER OC(C(C=C1)=CC=C1/C(C2=N/C(C=C2)=C3/C4=CC=C(C(O)=O)C=C4)=C(C=C/5)/N6C5=C(C7=CC=C(C(O)=O)C=C7)/C(C=C/8)=NC8=C(C9=CC=C(C(O)=O)C=C9)/C%10=CC=C3N%10[Fe]6Cl)=O
Specifikationer
Kemiskt namn eller material Fe(III) meso-Tetra(4-carboxyphenyl)porphine Chloride