Läs mer
Febuxostat 2-Butyl Isomer, TRC
Produktkod. 30408261
Change view
Kvantitet:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30408261 1 g
30400696 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30408261 Leverantör Toronto Research Chemicals Leverantörsnummer TRCF2290401G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Febuxostat 2-Butyl Isomer
CAS 1335202-59-9
Molekylformel C16H16N2O3S
Kvantitet 1 g
Synonym 2-(4-(sec-Butoxy)-3-cyanophenyl)-4-methylthiazole-5-carboxylic Acid
InChI formel InChI=1S/C16H16N2O3S/c1-4-9(2)21-13-6-5-11(7-12(13)8-17)15-18-10(3)14(22-15)16(19)20/h5-7,9H,4H2,1-3H3,(H,19,20)
LEDER COC(=O)C(C)c1cc(F)c(cc1)c2ccccc2
Molekylvikt (g/mol) 316.37
Formel vikt 316.0882