Läs mer
Febuxostat n-Butyl Isomer, TRC
Produktkod. 30180376
Change view
Kvantitet:
10 mg
100 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30180376 250 mg
30179812 10 mg
30179949 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30180376 Leverantör Toronto Research Chemicals Leverantörsnummer TRCF229035250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1657014-33-9
Molekylformel C16H16N2O3S
Molekylvikt (g/mol) 316.37
Synonym 2-(4-Butoxy-3-cyanophenyl)-4-methyl-5-thiazolecarboxylic Acid
LEDER OC(C(SC(C1=CC(C#N)=C(OCCCC)C=C1)=N2)=C2C)=O
Specifications
Kemiskt namn eller material Febuxostat n-Butyl Isomer
Kvantitet 250 mg
InChI formel InChI=1S/C16H16N2O3S/c1-3-4-7-21-13-6-5-11(8-12(13)9-17)15-18-10(2)14(22-15)16(19)20/h5-6,8H,3-4,7H2,1-2H3,(H,19,20)
Formel vikt 316.0882