missing translation for 'onlineSavingsMsg'
Learn More

FECH Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 14466-1-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16823124

  • 3313.85 SEK / 150µL
Estimated Shipment: 02-12-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The protein encoded by this gene is localized to the mitochondrion, where it catalyzes the insertion of the ferrous form of iron into protoporphyrin IX in the heme synthesis pathway. Mutations in this gene are associated with erythropoietic protoporphyria. Two transcript variants encoding different isoforms have been found for this gene. A pseudogene of this gene is found on chromosome 3.
Specifications

Specifications

FECH
0.18 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P22315, P22830
EPP, FCE, FECH, Ferrochelatase, mitochondrial, Heme synthase, Protoheme ferro lyase
Rabbit
IgG
150 μL
-20°C
Primary
14151, 2235, 361338
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
FECH
Liquid
Fech
FECH Fusion Protein Ag5814
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Videos
SDS
Documents
Certificates
Special Offers

Special Offers