Läs mer
Felbamate carbonate dimer, TRC
Produktkod. 30353571
Change view
Kvantitet:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30353571 100 mg
30363180 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30353571 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC177020100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1796927-91-7
Molekylformel C21H24N2O7
Molekylvikt (g/mol) 416.42
Synonym 3,3’-Carbonylbis(oxy)bis(2-phenylpropane-3,1-diyl) Dicarbamate, Felbamate Dimer
IUPAC-namn bis(3-carbamoyloxy-2-phenylpropyl) carbonate
LEDER NC(=O)OCC(COC(=O)OCC(COC(=O)N)c1ccccc1)c2ccccc2
Specifikationer
Kemiskt namn eller material 3,3'-Carbonylbis(oxy)bis(2-phenylpropane-3,1-diyl) Dicarbamate
Kvantitet 100 mg
InChI formel InChI=1S/C21H24N2O7/c22-19(24)27-11-17(15-7-3-1-4-8-15)13-29-21(26)30-14-18(12-28-20(23)25)16-9-5-2-6-10-16/h1-10,17-18H,11-14H2,(H2,22,24)(H2,23,25)
Formel vikt 416.1584