Läs mer
Felodipine Ester Lactone, TRC
Produktkod. 30404305
Change view
Kvantitet:
1 mg
Produktkod. Kvantitet
30404305 1 mg
1 options
Produktkod. 30404305 Leverantör Toronto Research Chemicals Leverantörsnummer TRCF2323901MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Felodipine Ester Lactone
CAS 85825-42-9
Molekylformel C17H15Cl2NO4
Kvantitet 1 mg
Synonym 4-(2,3-Dichlorophenyl)-1,4,5,7-tetrahydro-2-methyl-5-oxo-furo[3,4-b]pyridine-3-carboxylic Acid Ethyl Ester
InChI formel InChI=1S/C17H15Cl2NO4/c1-3-23-16(21)12-8(2)20-11-7-24-17(22)14(11)13(12)9-5-4-6-10(18)15(9)19/h4-6,13,20H,3,7H2,1-2H3
LEDER CCOC(=O)C1=C(C)NC2=C(C1c3cccc(Cl)c3Cl)C(=O)OC2
Molekylvikt (g/mol) 368.21
Formel vikt 367.0378