Läs mer
Felypressin, MedChemExpress
Produktkod. 16488308
Encompass_Preferred
Change view
Kvantitet:
2 mg
5 mg
10 mg
50 mg
100 mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
16488308 10 mg
16454197 50 mg
16420376 2 mg
16410748 5 mg
16408046 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Produktkod. 16488308 Leverantör Medchem Express Leverantörsnummer HYA0182
MedChemExpress Felypressin (PLV-2) is a non-catecholamine vasoconstrictor and a vasopressin 1 agonist. Felypressin is widely used in dental procedures.
Felypressin (PLV-2) is a non-catecholamine vasoconstrictor and a vasopressin 1 agonist. Felypressin is widely used in dental procedures.
Specifikationer
Kemiskt namn eller material Felypressin
CAS 56-59-7
Färg White
Anteckningar om renhetsgrad Research
Molekylformel C46H65N13O11S2
Kvantitet 10 mg
Synonym PLV-2
Löslighetsinformation DMSO : ≥ 10.4 mg/mL (10.00 mM)
LEDER O=C([C@H](CSSC[C@@H](C(N[C@H](C1=O)CC2=CC=CC=C2)=O)N)NC([C@@H](NC([C@@H](NC([C@](N1)([H])CC3=CC=CC=C3)=O)CCC(N)=O)=O)CC(N)=O)=O)N(CCC4)[C@@H]4C(N[C@@H](CCCCN)C(NCC(N)=O)=O)=O
Molekylvikt (g/mol) 1040.22
Formel vikt 1040.22
Kvalitet Research
Fysisk form Solid
För användning med (applikation) COVID-19-immunoregulation