Läs mer
Fenbuconazole-lactone A R-9129, TRC
Produktkod. 30453003
Change view
Kvantitet:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30453003 1 mg
30452034 5 mg
30455111 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30453003 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT7677251MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 146887-37-8
Molekylformel C19H16ClN3O2
Molekylvikt (g/mol) 353.8
Synonym Fenbuconazole-lactone A R-9129, trans-5-(4-Chlorophenyl)dihydro-3-phenyl-3-(1H-1,2,4-triazol-1-ylmethyl)-2(3H)-furanone, rel-(3R,5S)-5-(4-Chlorophenyl)dihydro-3-phenyl-3-(1H-1,2,4-triazol-1-ylmethyl)-2(3H)-furanone
IUPAC-namn (3S,5R)-5-(4-chlorophenyl)-3-phenyl-3-(1,2,4-triazol-1-ylmethyl)oxolan-2-one
LEDER Clc1ccc(cc1)[C@H]2C[C@](Cn3cncn3)(C(=O)O2)c4ccccc4
Specifikationer
Kemiskt namn eller material (3S,5R)-3-((1H-1,2,4-Triazol-1-yl)methyl)-5-(4-chlorophenyl)-3-phenyldihydrofuran-2(3H)-one
Kvantitet 1 mg
InChI formel InChI=1S/C19H16ClN3O2/c20-16-8-6-14(7-9-16)17-10-19(18(24)25-17,11-23-13-21-12-22-23)15-4-2-1-3-5-15/h1-9,12-13,17H,10-11H2/t17-,19-/m1/s1
Formel vikt 353.0931