Läs mer
Fenbufen, TRC
Produktkod. 30040732
Change view
Kvantitet:
10 mg
100 mg
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30040732 100 mg
18002620 25 g
18082120 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30040732 Leverantör Toronto Research Chemicals Leverantörsnummer TRCF247200100MG
Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!
This item requires a price update prior to being ordered.
Request a Price for this item here.
, 02 950 59 478
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 36330-85-5
Molekylformel C16 H14 O3
Molekylvikt (g/mol) 254.28
Synonym 3-(4-Biphenylcarbonyl)propionic acid, 3-(4-Phenylbenzoyl)propionic acid, 3-[(Biphenyl-4-yl)carbonyl]propionic acid, 4-(1,1'-Biphenyl-4-yl)-4-oxobutanoic acid, 4-(4-Phenylphenyl)-4-oxobutyric acid, 4-(Biphenyl-4-yl)-4-oxobutanoic acid, Bufemid, CL 82204, Cinopal, Cinopol, Fenbufen, Lederfen, Napanol, γ-Oxo[1,1'-biphenyl]-4-butanoic acid
IUPAC-namn 4-oxo-4-(4-phenylphenyl)butanoic acid
LEDER OC(=O)CCC(=O)c1ccc(cc1)c2ccccc2
Specifikationer
Kemiskt namn eller material Fenbufen
Kvantitet 100 mg
InChI formel InChI=1S/C16H14O3/c17-15(10-11-16(18)19)14-8-6-13(7-9-14)12-4-2-1-3-5-12/h1-9H,10-11H2,(H,18,19)
Formel vikt 254.0943