Läs mer
Fenbutrazate Hydrochloride, LoGiCal
Produktkod. 30462873
Change view
Kvantitet:
5 mg
Produktkod. Kvantitet
30462873 5 mg
1 options
Produktkod. 30462873 Leverantör LoGiCal Leverantörsnummer LGCFOR1759.00
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Fenbutrazate Hydrochloride
CAS 6474-85-7
Analyt- eller komponentnamn Fenbutrazate Hydrochloride
Frakt skick Room Temperature
Molekylformel C23 H29 N O3 . Cl H
Certifikat och efterlevnad ISO 9001
Synonym Fenbutrazate Hydrochloride, Benzeneacetic acid, α-ethyl-, 2-(3-methyl-2-phenyl-4-morpholinyl)ethyl ester, hydrochloride (1:1), Benzeneacetic acid, α-ethyl-, 2-(3-methyl-2-phenyl-4-morpholinyl)ethyl ester, hydrochloride (9CI), Butyric acid, 2-phenyl-, 2-(3-methyl-2-phenylmorpholino)ethyl ester, hydrochloride (6CI,7CI,8CI), Fenbutrazate hydrochloride, Phenbutrazate hydrochloride, R 381
InChI formel InChI=1S/C23H29NO3.ClH/c1-3-21(19-10-6-4-7-11-19)23(25)27-17-15-24-14-16-26-22(18(24)2)20-12-8-5-9-13-20;/h4-13,18,21-22H,3,14-17H2,1-2H3;1H
Lösningstyp Neat
LEDER Cl.CCC(C(=O)OCCN1CCOC(C1C)c2ccccc2)c3ccccc3
IUPAC-namn 2-(3-methyl-2-phenylmorpholin-4-yl)ethyl 2-phenylbutanoate;hydrochloride
Industry Type Clinical Testing
Molekylvikt (g/mol) 403.94
Kvantitet 5 mg
Formel vikt 403.1914 g/mol