Läs mer
(+)-Fenchone, TRC
Produktkod. 30407935
Change view
Kvantitet:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30407935 1 g
30397450 5 g
30403189 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30407935 Leverantör Toronto Research Chemicals Leverantörsnummer TRCF2474201G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 4695-62-9
Molekylformel C10 H16 O
Molekylvikt (g/mol) 152.23
Synonym Bicyclo[2.2.1]heptan-2-one, 1,3,3-trimethyl-, (1S,4R)-, 2-Norbornanone, 1,3,3-trimethyl-, (1S,4R)-(+)- (8CI), Bicyclo[2.2.1]heptan-2-one, 1,3,3-trimethyl-, (1S)-, (+)-Fenchone, (1S)-Fenchone, (1S,4R)-1,3,3-Trimethylbicyclo[2.2.1]heptan-2-one, (S)-(+)-Fenchone, (S)-Fenchone, D(+)-Fenchone, D-Fenchone
IUPAC-namn (1R,4S)-2,2,4-trimethylbicyclo[2.2.1]heptan-3-one
LEDER C[C@]12CC[C@H](C1)C(C)(C)C2=O
Specifikationer
Kemiskt namn eller material (+)-Fenchone
Kvantitet 1 g
InChI formel InChI=1S/C10H16O/c1-9(2)7-4-5-10(3,6-7)8(9)11/h7H,4-6H2,1-3H3/t7-,10+/m1/s1
Formel vikt 152.1201