Läs mer
Tocris Bioscience™ Fendiline Hydrochloride
GREENER_CHOICE
Produktkod. 18798158
Change view
Kvantitet:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
18798158 10 mg
18775949 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 18798158 Leverantör Tocris Bioscience™ Leverantörsnummer 6407/10
Inhibits KRas localization to the plasma membrane; also L-type calcium channel blocker
Fendiline Hydrochloride is a K-Ras inhibitor. Selectively inhibits K-Ras localization to the plasma membrane (IC50 = 9.64 μM). Blocks proliferation of cancer cell lines expressing oncogenic mutant K-Ras. Also inhibits L-type calcium channels (IC50 = 17 μM).
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Ras GTPase Inhibitors
CAS 13636-18-5
Analysprocentintervall >98%
Kvantitet 10 mg
Molekylformel C23H25N.HCl
Formel vikt Observed MW: 351.91