Läs mer
Fendiline hydrochloride, Thermo Scientific™
Brand: Thermo Scientific Alfa Aesar J63254.06
| Kvantitet | 5g |
|---|
Beskrivning
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
ApplicationsFendiline hydrochloride is used as a calcium channel blocker.
Solubility
Soluble in ethanol.
Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifikationer
| Fendiline hydrochloride | |
| C23H26ClN | |
| 5g | |
| fendiline hydrochloride, fendiline hcl, dsstox_cid_25860, dsstox_rid_81180, dsstox_gsid_45860, 3,3-diphenyl-n-1-phenylethyl propan-1-amine hydrochloride, prestwick_568, sensit tn | |
| NJXWZWXCHBNOOG-UHFFFAOYSA-N | |
| 3,3-diphenyl-N-(1-phenylethyl)propan-1-amine;hydrochloride | |
| 5702162 |
| 13636-18-5 | |
| MFCD00079301 | |
| 14,3970 | |
| Soluble in ethanol. | |
| CC(C1=CC=CC=C1)NCCC(C2=CC=CC=C2)C3=CC=CC=C3.Cl | |
| 351.918 | |
| 351.92 |
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.