Läs mer
Fenfluthrin, TRC
Produktkod. 30136981
Change view
Kvantitet:
100 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30136981 250 mg
18071220 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30136981 Leverantör Toronto Research Chemicals Leverantörsnummer TRCF247615250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 75867-00-4
Molekylformel C15 H11 Cl2 F5 O2
Molekylvikt (g/mol) 389.14
Synonym Cyclopropanecarboxylic acid, 3-(2,2-dichloroethenyl)-2,2-dimethyl-, (2,3,4,5,6-pentafluorophenyl)methyl ester, (1R,3S)-, Cyclopropanecarboxylic acid, 3-(2,2-dichloroethenyl)-2,2-dimethyl-, (pentafluorophenyl)methyl ester, (1R,3S)- (9CI), Cyclopropanecarboxylic acid, 3-(2,2-dichloroethenyl)-2,2-dimethyl-, (pentafluorophenyl)methyl ester, (1R-trans)-, Fenfluthrin, NAK 1654
IUPAC-namn (2,3,4,5,6-pentafluorophenyl)methyl (1R,3S)-3-(2,2-dichloroethenyl)-2,2-dimethylcyclopropane-1-carboxylate
LEDER CC1(C)[C@H](C=C(Cl)Cl)[C@H]1C(=O)OCc2c(F)c(F)c(F)c(F)c2F
Specifikationer
Kemiskt namn eller material Fenfluthrin
Anteckningar om renhetsgrad HPLC
Kvantitet 250 mg
InChI formel InChI=1S/C15H11Cl2F5O2/c1-15(2)6(3-7(16)17)8(15)14(23)24-4-5-9(18)11(20)13(22)12(21)10(5)19/h3,6,8H,4H2,1-2H3/t6-,8+/m1/s1
Formel vikt 388.0056
Procent renhet >95%