Beskrivning
Honeywell Fluka™ offers an expanding range of high quality inorganic certified reference materials and organic reference materials.
- Produced according to ISO 9001
- Suitable for a wide range of industries and applications
- Detailed Certificate of Analysis included with every delivery
Specifikationer
Specifikationer
| Kemiskt namn eller material | Fenhexamid |
| CAS | 126833-17-8 |
| CAS Max % | 1 |
| Densitet | 1.34 g/cm3 (20°C) |
| Flampunkt | >150°C |
| Beilstein | 8996658 |
| Färg | Beige |
| Smältpunkt | 155°C |
| Synonym | N-(2,3-Dichloro-4-hydroxyphenyl)-1-methylcyclohexanecarboxamide |
| Molekylvikt (g/mol) | 302.20 g/mol |
| Visa mer |